accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7789-39-1

BrRb

CAS RN 7789-39-1

IUPAC Name: rubidium(1+) bromide
Synonyms: rubidium bromide
Molecular Weight (g/mol): 165.37
Molecular Formula: BrRb
InChi Key: JAAGVIUFBAHDMA-UHFFFAOYSA-M
SMILES: [Br-].[Rb+]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)