CAS RN 7789-42-6
IUPAC Name:
cadmium(2+) dibromide
Synonyms:
cadmium bromide
Molecular Weight (g/mol):
272.22
Molecular Formula:
Br2Cd
InChi Key:
KPWJBEFBFLRCLH-UHFFFAOYSA-L
SMILES:
[Br-].[Br-].[Cd++]
1
–
3
of
3
results
1
Cadmium bromide, ultra dry, 99.999% (metals basis)
CAS: 7789-42-6 MDL Number: MFCD00015996 Synonym: cadmium 2+ dibromide,cadmium 2+ ion dibromide,cadmium bromide, anhydrous 25g,cadmium bromide, anhydrous,
2
Cadmium bromide, 98%, pure, anhydrous
CAS: 7789-42-6 Molecular Formula: Br2Cd Molecular Weight (g/mol): 272.22 Synonym: cadmium 2+ dibromide,cadmium 2+ ion dibromide,cadmium bromide, anhydrous 25g,cadmium bromide, anhydrous,
3
Cadmium bromide, anhydrous, 98%
CAS: 7789-42-6 Molecular Formula: CdBr2 MDL Number: MFCD00015996 Synonym: cadmium 2+ dibromide,cadmium 2+ ion dibromide,cadmium bromide, anhydrous 25g,cadmium bromide, anhydrous,