accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7789-43-7

CoBrBr

CAS RN 7789-43-7

IUPAC Name: λ²-cobalt(2+) dibromide
Synonyms: cobalt(II) bromide
Molecular Weight (g/mol): 218.74
Molecular Formula: Br2Co
InChi Key: BZRRQSJJPUGBAA-UHFFFAOYSA-L
SMILES: [Co++].[Br-].[Br-]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)