accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrBrHg

CAS RN 7789-47-1

IUPAC Name: mercury(2+) dibromide
Synonyms: mercury(II) bromide
Molecular Weight (g/mol): 360.4
Molecular Formula: Br2Hg
InChi Key: NGYIMTKLQULBOO-UHFFFAOYSA-L
SMILES: [Br-].[Br-].[Hg++]
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)