accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

MgBrBr

CAS RN 7789-48-2

IUPAC Name: magnesium(2+) dibromide
Synonyms: magnesium bromide
Molecular Weight (g/mol): 184.11
Molecular Formula: Br2Mg
InChi Key: OTCKOJUMXQWKQG-UHFFFAOYSA-L
SMILES: [Mg++].[Br-].[Br-]
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results