accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7789-60-8

IUPAC Name: tribromophosphane
Synonyms: phosphorus tribromide phosphorous tribromide
Molecular Weight (g/mol): 270.69
Molecular Formula: Br3P
InChi Key: IPNPIHIZVLFAFP-UHFFFAOYSA-N
SMILES: BrP(Br)Br
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (11)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (11)
  • (3)
  • (6)
  • (3)
  • (3)