CAS RN 7789-75-5
Calcium fluoride, 99%, pure, anhydrous
CAS: 7789-75-5 Molecular Formula: CaF2 Molecular Weight (g/mol): 78.08 MDL Number: MFCD00010907 InChI Key: WUKWITHWXAAZEY-UHFFFAOYSA-L Synonym: calcium fluoride,kalziumfluorid,kalziumdifluorid,calcium 2+ difluoride,acmc-1bk0c,ksc377c5h,calcium fluoride 100g,calcium fluoride,optical grade PubChem CID: 84512 ChEBI: CHEBI:35437 SMILES: [F-].[F-].[Ca++]
Calcium fluoride, 99%
CAS: 7789-75-5 Molecular Formula: CaF2 Molecular Weight (g/mol): 78.08 MDL Number: MFCD00010907 InChI Key: WUKWITHWXAAZEY-UHFFFAOYSA-L Synonym: calcium fluoride,kalziumfluorid,kalziumdifluorid,calcium 2+ difluoride,acmc-1bk0c,ksc377c5h,calcium fluoride 100g,calcium fluoride,optical grade PubChem CID: 84512 ChEBI: CHEBI:35437 SMILES: [F-].[F-].[Ca++]
Calcium fluoride, ultra dry, 99.99% (metals basis)
CAS: 7789-75-5 Molecular Formula: CaF2 Molecular Weight (g/mol): 78.08 MDL Number: MFCD00010907 InChI Key: WUKWITHWXAAZEY-UHFFFAOYSA-L Synonym: calcium fluoride,kalziumfluorid,kalziumdifluorid,calcium 2+ difluoride,acmc-1bk0c,ksc377c5h,calcium fluoride 100g,calcium fluoride,optical grade PubChem CID: 84512 ChEBI: CHEBI:35437 SMILES: [F-].[F-].[Ca++]
Calcium fluoride, 97%
CAS: 7789-75-5 Molecular Formula: CaF2 Molecular Weight (g/mol): 78.08 MDL Number: MFCD00010907 InChI Key: WUKWITHWXAAZEY-UHFFFAOYSA-L Synonym: calcium fluoride,kalziumfluorid,kalziumdifluorid,calcium 2+ difluoride,acmc-1bk0c,ksc377c5h,calcium fluoride 100g,calcium fluoride,optical grade PubChem CID: 84512 ChEBI: CHEBI:35437 IUPAC Name: calcium;difluoride SMILES: [F-].[F-].[Ca++]
Calcium fluoride, 99.5% (metals basis)
CAS: 7789-75-5 Molecular Formula: CaF2 Molecular Weight (g/mol): 78.08 MDL Number: MFCD00010907 InChI Key: WUKWITHWXAAZEY-UHFFFAOYSA-L Synonym: calcium fluoride,kalziumfluorid,kalziumdifluorid,calcium 2+ difluoride,acmc-1bk0c,ksc377c5h,calcium fluoride 100g,calcium fluoride,optical grade PubChem CID: 84512 ChEBI: CHEBI:35437 SMILES: [F-].[F-].[Ca++]
Calcium fluoride, Puratronic™, 99.985% (metals basis)
CAS: 7789-75-5 Molecular Formula: CaF2 Molecular Weight (g/mol): 78.08 MDL Number: MFCD00010907 InChI Key: WUKWITHWXAAZEY-UHFFFAOYSA-L Synonym: calcium fluoride,kalziumfluorid,kalziumdifluorid,calcium 2+ difluoride,acmc-1bk0c,ksc377c5h,calcium fluoride 100g,calcium fluoride,optical grade PubChem CID: 84512 ChEBI: CHEBI:35437 SMILES: [F-].[F-].[Ca++]
Calcium fluoride, Vacuum Deposition Grade, 99.9% (metals basis)
CAS: 7789-75-5 Molecular Formula: CaF2 Molecular Weight (g/mol): 78.08 MDL Number: MFCD00010907 InChI Key: WUKWITHWXAAZEY-UHFFFAOYSA-L Synonym: calcium fluoride,kalziumfluorid,kalziumdifluorid,calcium 2+ difluoride,acmc-1bk0c,ksc377c5h,calcium fluoride 100g,calcium fluoride,optical grade PubChem CID: 84512 ChEBI: CHEBI:35437 SMILES: [F-].[F-].[Ca++]
Calcium fluoride, 99%
CAS: 7789-75-5 Molecular Formula: CaF2 Molecular Weight (g/mol): 78.08 MDL Number: MFCD00010907 InChI Key: WUKWITHWXAAZEY-UHFFFAOYSA-L Synonym: calcium fluoride,kalziumfluorid,kalziumdifluorid,calcium 2+ difluoride,acmc-1bk0c,ksc377c5h,calcium fluoride 100g,calcium fluoride,optical grade PubChem CID: 84512 ChEBI: CHEBI:35437 IUPAC Name: calcium;difluoride SMILES: [F-].[F-].[Ca++]
Calcium fluoride, Reagent Grade, 97%
CAS: 7789-75-5 Molecular Formula: CaF2 Molecular Weight (g/mol): 78.08 MDL Number: MFCD00010907 InChI Key: WUKWITHWXAAZEY-UHFFFAOYSA-L Synonym: calcium fluoride,kalziumfluorid,kalziumdifluorid,calcium 2+ difluoride,acmc-1bk0c,ksc377c5h,calcium fluoride 100g,calcium fluoride,optical grade PubChem CID: 84512 ChEBI: CHEBI:35437 SMILES: [F-].[F-].[Ca++]
Calcium fluoride, 99.95% (metals basis)
CAS: 7789-75-5 Molecular Formula: CaF2 Molecular Weight (g/mol): 78.08 MDL Number: MFCD00010907 InChI Key: WUKWITHWXAAZEY-UHFFFAOYSA-L Synonym: calcium fluoride,kalziumfluorid,kalziumdifluorid,calcium 2+ difluoride,acmc-1bk0c,ksc377c5h,calcium fluoride 100g,calcium fluoride,optical grade PubChem CID: 84512 ChEBI: CHEBI:35437 SMILES: [F-].[F-].[Ca++]
Calcium Fluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Calcium Fluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Calcium Fluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
- 1
- 2