accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 779-89-5

IUPAC Name: (2E)-3-[3-(trifluoromethyl)phenyl]prop-2-enoate
Synonyms: (2E)-3-[3-(trifluoromethyl)phenyl]prop-2-enoate
Molecular Weight (g/mol): 215.15
Molecular Formula: C10H6F3O2
InChi Key: KSBWHDDGWSYETA-SNAWJCMRSA-M
SMILES: [O-]C(=O)\C=C\C1=CC=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results