accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7790-30-9

ITl

CAS RN 7790-30-9

IUPAC Name: λ¹-thallanylium iodide
Synonyms: thallium(I) iodide
Molecular Weight (g/mol): 331.28
Molecular Formula: ITl
InChi Key: CMJCEVKJYRZMIA-UHFFFAOYSA-M
SMILES: [I-].[Tl+]
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)