accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7790-47-8

SnIHIHIHHI

CAS RN 7790-47-8

IUPAC Name: tin(IV) iodide
Synonyms: tin(IV) iodide
Molecular Weight (g/mol): 630.36
Molecular Formula: H4I4Sn
InChi Key: ZBCVLXKKLMNLGV-UHFFFAOYSA-N
SMILES: [Sn].I.I.I.I
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)