accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7790-75-2

CaOWOOO

CAS RN 7790-75-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 287.91
Molecular Formula: CaO4W
InChi Key: FDMFQOCGNBYKPY-UHFFFAOYSA-N
SMILES: [Ca++].[O-][W]([O-])(=O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)