accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7790-80-9

CdII

CAS RN 7790-80-9

IUPAC Name: cadmium(2+) diiodide
Synonyms: cadmium iodide
Molecular Weight (g/mol): 366.22
Molecular Formula: CdI2
InChi Key: OKIIEJOIXGHUKX-UHFFFAOYSA-L
SMILES: [Cd++].[I-].[I-]
Molecular Weight (g/mol) 
  • (1)
  • (10)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (9)
  • (1)
  • (10)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)