CAS RN 7790-84-3
CAS RN 7790-84-3
IUPAC Name:
cadmium(2+) hydrate sulfate
Synonyms:
cadmium(2+) hydrate sulfate
Molecular Weight (g/mol):
226.49
Molecular Formula:
CdH2O5S
InChi Key:
VRFRVKRTIMIRNG-UHFFFAOYSA-L
SMILES:
O.[Cd++].[O-]S([O-])(=O)=O
1
–
7
of
7
results
1
Cadmium sulfate, 8/3-hydrate, ACS reagent, crystals
CAS: 7790-84-3 Molecular Formula: Cd3O12S3·8H2O Molecular Weight (g/mol): 769.51 MDL Number: MFCD02253000
2
Cadmium sulfate octahydrate, ACS, 98.0-102.0%
CAS: 7790-84-3 MDL Number: MFCD00151655
3
Cadmium Sulfate, Extra Pure, SLR, Fisher Chemical™
CAS: 7790-84-3 Molecular Formula: CdH2O5S Molecular Weight (g/mol): 226.49 MDL Number: 2253000 InChI Key: VRFRVKRTIMIRNG-UHFFFAOYSA-L IUPAC Name: cadmium(2+) hydrate sulfate SMILES: O.[Cd++].[O-]S([O-])(=O)=O
5
Cadmium Sulfate 8/3-Hydrate, Honeywell Fluka™
CAS: 7790-84-3 MDL Number: MFCD02253000
6
Cadmium Sulfate 8/3-Hydrate, puriss. p.a. ACS Reagent, ≥99.0%, Honeywell Fluka™
CAS: 7790-84-3 MDL Number: MFCD02253000
7
Cadmium Sulfate, Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 7790-84-3 Molecular Formula: CdSO4.8/3 H2O