accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7791-12-0

IUPAC Name: λ¹-thallanylium chloride
Synonyms: thallium(I) chloride
Molecular Weight (g/mol): 239.83
Molecular Formula: ClTl
InChi Key: GBECUEIQVRDUKB-UHFFFAOYSA-M
SMILES: [Cl-].[Tl+]
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)