CAS RN 7791-20-0
CAS RN 7791-20-0
Nickel(II) chloride hexahydrate, 97%
CAS: 7791-20-0 Molecular Formula: Cl2Ni·6H2O Molecular Weight (g/mol): 237.71 MDL Number: MFCD00149809
Nickel(II) chloride hexahydrate, 99.95% (metals basis)
CAS: 7791-20-0 Molecular Formula: Cl2Ni·6H2O MDL Number: MFCD00149809
Nickel(II) chloride hexahydrate, 98%
CAS: 7791-20-0 Molecular Formula: Cl2Ni·6H2O MDL Number: MFCD00149809
Nickel(II) chloride hexahydrate, 98+%
CAS: 7791-20-0 Molecular Formula: Cl2Ni·6H2O Molecular Weight (g/mol): 237.71
Nickel(II) chloride hexahydrate, Puratronic™, 99.999% (metals basis)
CAS: 7791-20-0 Molecular Formula: Cl2Ni·6H2O MDL Number: MFCD00149809
Nickel(II) chloride hexahydrate, 99.3% (metals basis)
CAS: 7791-20-0 Molecular Formula: Cl2Ni·6H2O MDL Number: MFCD00149809
Nickel(II) chloride hexahydrate, 99.999%, (trace metal basis)
CAS: 7791-20-0 Molecular Formula: Cl2Ni·6H2O Molecular Weight (g/mol): 237.71 MDL Number: MFCD00149809
Nickel Chloride Hexahydrate, (Crystalline),MP Biomedicals
CAS: 7791-20-0 Molecular Formula: NiCl2 · 6H2O
Nickel Chloride Hexahydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Nickel Chloride Hexahydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Nickel(II) Chloride Hexahydrate, puriss. p.a., ≥98%, Honeywell Fluka™
CAS: 7791-20-0 MDL Number: MFCD00149809
Nickel Chloride Hexahydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Nickel(II) Chloride Hexahydrate, Certified AR for Analysis, Fisher Chemical™
CAS: 7791-20-0 Molecular Formula: Cl2Ni·6H2O MDL Number: 149809
Nickel(II) chloride, 0.5M aq. soln., Thermo Scientific™
CAS: 7791-20-0 Molecular Formula: Cl2Ni MDL Number: MFCD00011142
- 1
- 2