accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7791-25-5

IUPAC Name: sulfuroyl dichloride
Synonyms: sulfonyl chloride sulfurylchlorid sulfuryl chloride sulphuryl dichloride scl2O2 sulfuryl dichloride sulfuric dichloride SO2cl2 sulfuric oxychloride sulfur dichloride dioxide
Molecular Weight (g/mol): 134.96
Molecular Formula: Cl2O2S
InChi Key: YBBRCQOCSYXUOC-UHFFFAOYSA-N
SMILES: ClS(Cl)(=O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (2)