accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 77978-73-5

IUPAC Name: 2-[4-(methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid
Synonyms: [4-(methoxycarbonyl)-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl]acetic acid
Molecular Weight (g/mol): 213.19
Molecular Formula: C9H11NO5
InChi Key: WLKUVVIAHUXPSB-UHFFFAOYNA-N
SMILES: COC(=O)C1=C(C)NC(=O)C1CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results