accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSiCH3CH3OH3C

CAS RN 78-62-6

IUPAC Name: diethoxydimethylsilane
Synonyms: dimethyldiethoxysilane dimethyl diethoxysilane
Molecular Weight (g/mol): 148.28
Molecular Formula: C6H16O2Si
InChi Key: YYLGKUPAFFKGRQ-UHFFFAOYSA-N
SMILES: CCO[Si](C)(C)OCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results