accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78-70-6

IUPAC Name: 3,7-dimethylocta-1,6-dien-3-ol
Synonyms: linalool, (+-)- epa pesticide chemical code 128838 (s)-linalool caswell no. 526A fema number 2635 linalool (natural) (S)-isomer linalool linalool, (R)-isomer p-linalool (s)-isomer linalool
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: CDOSHBSSFJOMGT-UHFFFAOYNA-N
SMILES: CC(C)=CCCC(C)(O)C=C
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)