accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3BrBr

CAS RN 78-75-1

IUPAC Name: 1,2-dibromopropane
Synonyms: 1,2-dibromopropane
Molecular Weight (g/mol): 201.89
Molecular Formula: C3H6Br2
InChi Key: XFNJYAKDBJUJAJ-UHFFFAOYNA-N
SMILES: CC(Br)CBr
Concentration 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results