accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78-78-4

IUPAC Name: 2-methylbutane
Synonyms: isopentane C7-8 isoparaffin iso-C5H12
Molecular Weight (g/mol): 72.15
Molecular Formula: C5H12
InChi Key: QWTDNUCVQCZILF-UHFFFAOYSA-N
SMILES: CCC(C)C
Quantity 
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (6)
Grade 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (8)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)