accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CH2N

CAS RN 78-81-9

IUPAC Name: 2-methylpropan-1-amine
Synonyms: isobutylamine valamine I-butylamine 3-methyl-2-propylamine iso-C4H9NH2
Molecular Weight (g/mol): 73.14
Molecular Formula: C4H11N
InChi Key: KDSNLYIMUZNERS-UHFFFAOYSA-N
SMILES: CC(C)CN
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)