accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78-86-4

IUPAC Name: 2-chlorobutane
Synonyms: 2-chlorobutane
Molecular Weight (g/mol): 92.57
Molecular Formula: C4H9Cl
InChi Key: BSPCSKHALVHRSR-UHFFFAOYNA-N
SMILES: CCC(C)Cl
Boiling Point 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)