accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7803-57-8

H2ONH2H2N

CAS RN 7803-57-8

IUPAC Name: hydrazine hydrate
Synonyms: hydrazine hydrate hydrazinium hydroxide hydrazine hydroxide
Molecular Weight (g/mol): 50.06
Molecular Formula: H6N2O
InChi Key: IKDUDTNKRLTJSI-UHFFFAOYSA-N
SMILES: O.NN
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (4)
  • (4)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (9)
  • (1)