accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SNH2OO

CAS RN 7803-58-9

IUPAC Name: sulfamoylamine
Synonyms: sulfamide sulfonyl diamide imidosulfamic acid sulfuryl diamide diamidodioxidosulfur sulfamamide sulfuryl amide
Molecular Weight (g/mol): 96.1
Molecular Formula: H4N2O2S
InChi Key: NVBFHJWHLNUMCV-UHFFFAOYSA-N
SMILES: NS(N)(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)