accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78056-39-0

IUPAC Name: 4,5-difluoro-2-nitroaniline
Synonyms: 4,5-difluoro-2-nitroaniline
Molecular Weight (g/mol): 174.11
Molecular Formula: C6H4F2N2O2
InChi Key: WDMCABATCGQAKK-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(F)C=C1[N+]([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results