accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(S)(S)NHONOCH3H3CHOOSNH3NONSOOO(Z)

CAS RN 78110-38-0

IUPAC Name: (2S,3S)-3-[(2Z)-2-(2-azaniumyl-1,3-thiazol-4-yl)-2-[(1-carboxy-1-methylethoxy)imino]acetamido]-2-methyl-4-oxoazetidine-1-sulfonate
Synonyms: aztreonam
Molecular Weight (g/mol): 435.43
Molecular Formula: C13H17N5O8S2
InChi Key: WZPBZJONDBGPKJ-VEHQQRBSSA-N
SMILES: C[C@H]1[C@H](NC(=O)C(=N/OC(C)(C)C(O)=O)\C2=CSC([NH3+])=N2)C(=O)N1S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (4)
  • (3)
  • (1)
  • (2)
Type 
  • (6)
Percent Purity 
  • (2)
  • (3)
Color 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (7)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (6)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)