accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3SOONN

CAS RN 78162-58-0

IUPAC Name: N,N-dimethyl-1H-imidazole-1-sulfonamide
Synonyms: N,N-dimethylimidazole-1-sulfonamide
Molecular Weight (g/mol): 175.21
Molecular Formula: C5H9N3O2S
InChi Key: YRRWNBMOJMMXQY-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)N1C=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results