CAS RN 78281-72-8
IUPAC Name:
2-(2-amino-3-benzoylphenyl)acetamide
Synonyms:
nepafenac amfenac amide nevanac
Molecular Weight (g/mol):
254.29
Molecular Formula:
C15H14N2O2
InChi Key:
QEFAQIPZVLVERP-UHFFFAOYSA-N
SMILES:
NC(=O)CC1=CC=CC(C(=O)C2=CC=CC=C2)=C1N
1
–
3
of
3
results
1
Nepafenac, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Nepafenac, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Nepafenac, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.