accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78287-27-1

IUPAC Name: (19S)-10,19-diethyl-19-hydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Synonyms: (19S)-10,19-diethyl-19-hydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Molecular Weight (g/mol): 376.41
Molecular Formula: C22H20N2O4
InChi Key: MYQKIWCVEPUPIL-QFIPXVFZSA-N
SMILES: CCC1=C2C=CC=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)