accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONNH2FFF

CAS RN 78335-26-9

IUPAC Name: 4-(morpholin-4-yl)-2-(trifluoromethyl)aniline
Synonyms: 4-(morpholin-4-yl)-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 246.23
Molecular Formula: C11H13F3N2O
InChi Key: DZEZKCZKDBIQSW-UHFFFAOYSA-N
SMILES: NC1=C(C=C(C=C1)N1CCOCC1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results