accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3NNOH3COCH3

CAS RN 78342-42-4

IUPAC Name: 3,6-dimethoxy-2-(propan-2-yl)-2,5-dihydropyrazine
Synonyms: 3-isopropyl-2,5-dimethoxy-3,6-dihydropyrazine
Molecular Weight (g/mol): 184.24
Molecular Formula: C9H16N2O2
InChi Key: FCFWEOGTZZPCTO-UHFFFAOYNA-N
SMILES: COC1=NC(C(C)C)C(OC)=NC1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results