accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 784-50-9

IUPAC Name: 9-oxo-9H-fluorene-2-carboxylic acid
Synonyms: 9-oxofluorene-2-carboxylic acid
Molecular Weight (g/mol): 224.22
Molecular Formula: C14H8O3
InChi Key: BJCTXUUKONLPPK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C3=CC=CC=C3C(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results