accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 784-71-4

IUPAC Name: 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 246.19
Molecular Formula: C9H11FN2O5
InChi Key: UIYWFOZZIZEEKJ-XVFCMESISA-N
SMILES: OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results