accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OCH3H3CONONH(R)(R)SNOOONHCSNH2NHOO(Z)

CAS RN 78439-06-2

IUPAC Name: 1-{[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(1-carboxy-1-methylethoxy)imino]acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}-1,2-dihydropyridin-2-ylium pentahydrate
Synonyms: 1-{[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(1-carboxy-1-methylethoxy)imino]acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}-2H-pyridin-2-ylium pentahydrate
Molecular Weight (g/mol): 636.65
Molecular Formula: C22H32N6O12S2
InChi Key: NMVPEQXCMGEDNH-TZVUEUGBSA-N
SMILES: O.O.O.O.O.CC(C)(O\N=C(/C(=O)N[C@H]1[C@H]2SCC(CN3[CH+]C=CC=C3)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)