accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78613-38-4

IUPAC Name: hydrogen (2R,6S)-2,6-dimethyl-4-{2-methyl-3-[4-(2-methylbutan-2-yl)phenyl]propyl}morpholine chloride
Synonyms: hydrogen amorolfine chloride
Molecular Weight (g/mol): 353.98
Molecular Formula: C21H36ClNO
InChi Key: XZKWIPVTHGWDCF-KUZYQSSXSA-N
SMILES: [H+].[Cl-].CCC(C)(C)C1=CC=C(CC(C)CN2C[C@H](C)O[C@H](C)C2)C=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results