accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78628-80-5

IUPAC Name: hydrogen [(2E)-6,6-dimethylhept-2-en-4-yn-1-yl](methyl)[(naphthalen-1-yl)methyl]amine chloride
Synonyms: hydrogen terbinafine chloride
Molecular Weight (g/mol): 327.9
Molecular Formula: C21H26ClN
InChi Key: BWMISRWJRUSYEX-SZKNIZGXSA-N
SMILES: [H+].[Cl-].CN(C\C=C\C#CC(C)(C)C)CC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)