CAS RN 787-70-2
IUPAC Name:
[1,1'-biphenyl]-4,4'-dicarboxylic acid
Synonyms:
santalum album santalum album wood powder santalum album wood oil santalum album extract santalum album seed extract santalum album wood extract
Molecular Weight (g/mol):
242.23
Molecular Formula:
C14H10O4
InChi Key:
NEQFBGHQPUXOFH-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O
1
–
1
of
1
results
1
Promotions Available
4,4'-Biphenyldicarboxylic acid, 98%
4,4'-Biphenyldicarboxylic acid, 98%
CAS: 787-70-2 Molecular Formula: C14H10O4 Molecular Weight (g/mol): 242.23 MDL Number: MFCD00002554 InChI Key: NEQFBGHQPUXOFH-UHFFFAOYSA-N Synonym: biphenyl-4,4'-dicarboxylic acid,4,4'-biphenyldicarboxylic acid,1,1'-biphenyl-4,4'-dicarboxylic acid,4-4-carboxyphenyl benzoic acid,4,4'-bibenzoic acid,4,4'-diphenic acid,4,4'-dicarboxybiphenyl,p,p'-diphenic acid,usaf do-69,bibenzoic acid PubChem CID: 13084 IUPAC Name: 4-(4-carboxyphenyl)benzoic acid SMILES: C1=CC(=CC=C1C2=CC=C(C=C2)C(=O)O)C(=O)O