accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CClO

CAS RN 79-03-8

IUPAC Name: propanoyl chloride
Synonyms: propanoyl chloride
Molecular Weight (g/mol): 92.52
Molecular Formula: C3H5ClO
InChi Key: RZWZRACFZGVKFM-UHFFFAOYSA-N
SMILES: CCC(Cl)=O
Boiling Point 
  • (3)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (7)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (6)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)