accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79-10-7

IUPAC Name: prop-2-enoic acid
Synonyms: acrylic acid carbomer 1342 acrylic acid, fe (3+) salt acrylic acid, glacial ca (2:1) salt acrylic acid silver salt acrylic acid acroleic acid carbopol 1342 carbopol 934P sodium salt acrylic acid
Molecular Weight (g/mol): 72.06
Molecular Formula: C3H4O2
InChi Key: NIXOWILDQLNWCW-UHFFFAOYSA-N
SMILES: OC(=O)C=C
Molecular Weight (g/mol) 
  • (7)
  • (7)
Percent Purity 
  • (3)
  • (2)
  • (7)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (7)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (7)
  • (3)
  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (4)
  • (8)
  • (4)
  • (2)