accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOHO

CAS RN 79-21-0

IUPAC Name: ethaneperoxoic acid
Synonyms: peracetic acid peroxyethanoic acid peracetic acid (product) caswell no. 644 desoxone 1 desoxone-1 peroxoacetic acid peroxyacetic acid (substance) osbon AC ethaneperoxoic acid
Molecular Weight (g/mol): 76.05
Molecular Formula: C2H4O3
InChi Key: KFSLWBXXFJQRDL-UHFFFAOYSA-N
SMILES: CC(=O)OO
Molecular Weight (g/mol) 
  • (2)
  • (7)
Quantity 
  • (3)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results