accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COClO

CAS RN 79-22-1

IUPAC Name: methyl carbonochloridate
Synonyms: methyl chloroformate
Molecular Weight (g/mol): 94.49
Molecular Formula: C2H3ClO2
InChi Key: XMJHPCRAQCTCFT-UHFFFAOYSA-N
SMILES: COC(Cl)=O
Boiling Point 
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)