CAS RN 79-29-8
IUPAC Name:
2,3-dimethylbutane
Synonyms:
2,3-dimethylbutane
Molecular Weight (g/mol):
86.18
Molecular Formula:
C6H14
InChi Key:
ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
SMILES:
CC(C)C(C)C
1
–
3
of
3
results
1
2,3-Dimethylbutane, 98+%
CAS: 79-29-8 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00008925 InChI Key: ZFFMLCVRJBZUDZ-UHFFFAOYSA-N Synonym: diisopropyl,biisopropyl,butane, 2,3-dimethyl,1,1,2,2-tetramethylethane,hsdb 76,unii-68isq7a432,ccris 6020,ch3 2chch ch3 2,acmc-1bmfq,dsstox_cid_5112 PubChem CID: 6589 IUPAC Name: 2,3-dimethylbutane SMILES: CC(C)C(C)C
3
ASTM Method D5191 Vapor Pressure - 51.1kPa (7.41 psi), Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality