accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79-31-2

IUPAC Name: 2-methylpropanoic acid
Synonyms: isobutyric acid hemiammoniate isobutyric acid 2-methylpropionsaeure cenex RP b2 iso-C3H7COOH sobutyric acid nickel salt isobutyric acid ammonium salt isobutyric acid isobutyric acid (natural) isobuttersaeure
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: KQNPFQTWMSNSAP-UHFFFAOYSA-N
SMILES: CC(C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)