accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79-34-5

IUPAC Name: 1,1,2,2-tetrachloroethane
Synonyms: 1,1,2,2-tetrachloroethane caswell no. 826 tce (ambiguous) bonoform 1,1,2,2-tetrachloraethan sym-tetrachloroethane cellon acetylene tetrachloride 1,1,2,2-tetrachloorethaan 1,1,2,2-czterochloroetan
Molecular Weight (g/mol): 167.84
Molecular Formula: C2H2Cl4
InChi Key: QPFMBZIOSGYJDE-UHFFFAOYSA-N
SMILES: ClC(Cl)C(Cl)Cl
Molecular Weight (g/mol) 
  • (10)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (10)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (4)
  • (7)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)