accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOClO

CAS RN 79-37-8

IUPAC Name: oxalic dichloride
Synonyms: oxalyl chloride
Molecular Weight (g/mol): 126.92
Molecular Formula: C2Cl2O2
InChi Key: CTSLXHKWHWQRSH-UHFFFAOYSA-N
SMILES: ClC(=O)C(Cl)=O
Molecular Weight (g/mol) 
  • (6)
  • (9)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (10)
Boiling Point 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (9)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (10)
  • (4)
  • (4)