accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH2OHO

CAS RN 79-41-4

IUPAC Name: 2-methylprop-2-enoic acid
Synonyms: methacrylic acid 2-methylpropensaeure methakrylsaeure methacrylsaeure acido metacrilico acide methacrylique kyselina methakrylova
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: CERQOIWHTDAKMF-UHFFFAOYSA-N
SMILES: CC(=C)C(O)=O
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (7)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (3)
  • (3)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (4)
  • (9)
  • (6)