CAS RN 79-93-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phenaglycodol, TRC
CAS: 79-93-6 Molecular Formula: C11H15ClO2 Molecular Weight (g/mol): 214.69 Synonym: 2-(4-Chlorophenyl)-3-methyl-2,3-butanediol,2-p-Chlorophenyl-3-methyl-2,3-butanediol,Acalmid,Acalo,Alterton,Felixyn,NSC 169451,NSC 63894,Sinforil,Stesil,Ultran IUPAC Name: 2-(4-chlorophenyl)-3-methylbutane-2,3-diol SMILES: CC(C)(O)C(C)(O)c1ccc(Cl)cc1