accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79079-18-8

OOOOOOOOOOOOLaLa

CAS RN 79079-18-8

IUPAC Name: tris(oxalic acid) decahydrate dilanthanum
Synonyms: tris(oxalic acid) decahydrate dilanthanum
Molecular Weight (g/mol): 728.06
Molecular Formula: C6H26La2O22
InChi Key: SFCDNUVGLFIDSK-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.O.O.O.[La].[La].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results