accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 791-28-6

IUPAC Name: (diphenylphosphoroso)benzene
Synonyms: triphenylphosphine oxide triphenylphosphanoxid triphenyl phosphorus oxide triphenyl phosphine oxide
Molecular Weight (g/mol): 278.29
Molecular Formula: C18H15OP
InChi Key: FIQMHBFVRAXMOP-UHFFFAOYSA-N
SMILES: O=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (9)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (6)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results